Assay Design



Gene Transcripts

Gene Symbol
cyp2aa1
Entrez Gene ID
572000
Gene Name
cytochrome P450, family 2, subfamily AA, polypeptide 1
Gene Aliases
zgc:162502
Chromosome Location
Chr.23: 42434348 - 42451859 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025557.2 NP_001020728.2 7 - 7 1147 115
GenBank mRNA BC134006.1 - 7 - 7 1147 115

View Full Product Details