Assay Design



Gene Transcripts

Gene Symbol
PTGS2
Entrez Gene ID
791253
Gene Name
prostaglandin-endoperoxide synthase 2
Gene Aliases
COX-2, PGH2S, PGHS-2, PGHS2
Chromosome Location
Chr.5: 20490104 - 20497088 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081775.2 NP_001075244.1 3 - 4 432 70
GenBank mRNA AF027334.1 - 3 - 4 432 70

View Full Product Details