Assay Design



Gene Transcripts

Gene Symbol
MSTN
Entrez Gene ID
100033832
Gene Name
myostatin
Gene Aliases
-
Chromosome Location
Chr.18: 66605150 - 66610122 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081817.1 NP_001075286.1 2 - 3 742 93
GenBank mRNA AB033541.1 - 2 - 3 742 93

View Full Product Details