Assay Design



Gene Transcripts

Gene Symbol
AKR1C1
Entrez Gene ID
100034073
Gene Name
aldo-keto reductase family 1, member C1
Gene Aliases
AKR1C23
Chromosome Location
Chr.29: 30127645 - 30166814 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081908.1 NP_001075377.1 2 - 3 263 57
GenBank mRNA AY955082.1 - 2 - 3 263 57

View Full Product Details