Assay Design



Gene Transcripts

Gene Symbol
TIMP1
Entrez Gene ID
100034220
Gene Name
TIMP metallopeptidase inhibitor 1
Gene Aliases
TIMP-1
Chromosome Location
Chr.X: 40032651 - 40036523 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082515.1 NP_001075984.1 3 - 4 348 57
XM_023633181.1 XP_023488949.1 - 497 57
GenBank mRNA U95039.1 - 3 - 4 348 57

View Full Product Details