Assay Design



Gene Transcripts

Gene Symbol
IL23A
Entrez Gene ID
100034230
Gene Name
interleukin 23 subunit alpha
Gene Aliases
IL-23, IL23
Chromosome Location
Chr.6: 74880216 - 74885877 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082522.2 NP_001075991.1 2 - 3 341 63
XM_014740754.2 XP_014596240.1 - 1579 63
GenBank mRNA EU438773.1 - 2 - 3 341 63

View Full Product Details