Assay Design



Gene Transcripts

Gene Symbol
TLR8
Entrez Gene ID
100054367
Gene Name
toll like receptor 8
Gene Aliases
-
Chromosome Location
Chr.X: 9607530 - 9626631 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111301.1 NP_001104771.1 1 - 2 198 90
XM_005613949.3 XP_005614006.1 - 397 90
XM_014728737.2 XP_014584223.1 - 244 90
GenBank mRNA EF090622.1 - 1 - 2 198 90

View Full Product Details