Assay Design



Gene Transcripts

Gene Symbol
PPP6R3
Entrez Gene ID
100053490
Gene Name
protein phosphatase 6 regulatory subunit 3
Gene Aliases
SAPS3
Chromosome Location
Chr.12: 31467205 - 31607697 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113235.1 NP_001106706.1 15 - 16 1884 67
XM_023653807.1 XP_023509575.1 - 1916 67
XM_023653808.1 XP_023509576.1 - 1740 67
XM_023653810.1 XP_023509578.1 - 1748 67
XM_023653822.1 XP_023509590.1 - 1437 67
XM_023653823.1 XP_023509591.1 - 1496 67
XM_023653824.1 XP_023509592.1 - 1916 67
GenBank mRNA AB371895.1 - 15 - 16 1884 67

View Full Product Details