Assay Design



Gene Transcripts

Gene Symbol
GNS
Entrez Gene ID
100034131
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Gene Aliases
G6S
Chromosome Location
Chr.6: 81638600 - 81675726 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001492151.6 XP_001492201.6 - 1352 95
XM_023643942.1 XP_023499710.1 - 1352 95
GenBank mRNA AF510336.1 - 10 - 11 1240 95

View Full Product Details