Assay Design



Gene Transcripts

Gene Symbol
PITPNC1
Entrez Gene ID
100034047
Gene Name
phosphatidylinositol transfer protein cytoplasmic 1
Gene Aliases
-
Chromosome Location
Chr.11: 14270586 - 14515454 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163878.1 NP_001157350.1 6 - 7 1149 74
XM_005597019.3 XP_005597076.1 - 1340 74
XM_023651592.1 XP_023507360.1 - 1401 74
XM_023651593.1 XP_023507361.1 - 548 74
XM_023651594.1 XP_023507362.1 - 476 74
XM_023651596.1 XP_023507364.1 - 1400 74
GenBank mRNA AY500592.1 - 5 - 6 333 74

View Full Product Details