Assay Design



Gene Transcripts

Gene Symbol
EDN3
Entrez Gene ID
100034064
Gene Name
endothelin 3
Gene Aliases
-
Chromosome Location
Chr.22: 46643067 - 46665984 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_023626834.1 XP_023482602.1 - 987 118
GenBank mRNA AY762637.1 - 4 - 5 554 118

View Full Product Details