Assay Design



Gene Transcripts

Gene Symbol
TCRB
Entrez Gene ID
100054892
Gene Name
T-cell receptor beta chain
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY927792.1 - 1 - 2 97 71
AY927798.1 - 1 - 2 100 71
AY927805.1 - 1 - 2 68 71

View Full Product Details