Assay Design



Gene Transcripts

Gene Symbol
SELENOW
Entrez Gene ID
100050195
Gene Name
selenoprotein W
Gene Aliases
SEPW1
Chromosome Location
Chr.10: 18003671 - 18008557 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170422.2 NP_001163893.1 1 - 2 64 80

View Full Product Details