Assay Design



Gene Transcripts

Gene Symbol
HNRNPLL
Entrez Gene ID
416702
Gene Name
heterogeneous nuclear ribonucleoprotein L like
Gene Aliases
HNRPLL
Chromosome Location
Chr.3: 16581734 - 16608471 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305651.2 NP_001292580.2 11 - 12 1606 120
XM_015283458.2 XP_015138944.1 11 - 12 1556 120
GenBank mRNA BX929305.1 - 2 - 3 219 120
BX934454.1 - 4 - 5 554 120
CR407217.1 - 4 - 5 444 120

View Full Product Details