Assay Design



Gene Transcripts

Gene Symbol
MRPL32
Entrez Gene ID
420773
Gene Name
mitochondrial ribosomal protein L32
Gene Aliases
-
Chromosome Location
Chr.2: 51155614 - 51158036 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_418869.6 XP_418869.1 2 - 3 332 87
GenBank mRNA BX929537.1 - 2 - 3 289 87
BX930780.1 - 1 - 2 400 87
BX934915.1 - 2 - 3 327 87

View Full Product Details