Assay Design



Gene Transcripts

Gene Symbol
CDKN3
Entrez Gene ID
423562
Gene Name
cyclin dependent kinase inhibitor 3
Gene Aliases
-
Chromosome Location
Chr.5: 56665665 - 56671678 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252162.1 NP_001239091.1 5 - 6 553 98
GenBank mRNA BX929389.2 - 2 - 3 685 98
BX929945.1 - 5 - 6 347 98
BX933305.1 - 4 - 5 472 98
CR353075.1 - 4 - 5 422 98

View Full Product Details