Assay Design



Gene Transcripts

Gene Symbol
MICU3
Entrez Gene ID
422736
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
Chromosome Location
Chr.4: 63212802 - 63266757 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015285646.2 XP_015141132.1 7 - 8 1086 149
GenBank mRNA CR338835.1 - 5 - 6 629 149

View Full Product Details