Assay Design



Gene Transcripts

Gene Symbol
FAM173A
Entrez Gene ID
416404
Gene Name
family with sequence similarity 173 member A
Gene Aliases
C16orf24
Chromosome Location
Chr.14: 13580817 - 13582729 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277151.1 NP_001264080.1 2 - 3 283 116
NM_001277152.1 NP_001264081.1 2 - 3 283 116
NM_001277162.1 NP_001264091.1 2 - 3 283 116
XM_015294663.2 XP_015150149.1 2 - 3 287 116
GenBank mRNA BU241996.1 - 2 - 3 283 116
BX930158.1 - 2 - 3 273 116
BX930472.1 - 2 - 3 276 116
BX931208.2 - 2 - 3 272 116
BX950531.1 - 2 - 3 279 116

View Full Product Details