Assay Design



Gene Transcripts

Gene Symbol
METTL7A
Entrez Gene ID
426871
Gene Name
methyltransferase like 7A
Gene Aliases
-
Chromosome Location
Chr.33: 3166735 - 3168614 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_424479.6 XP_424479.3 1 - 2 697 75
GenBank mRNA BX929440.2 - 1 - 2 686 75
BX931252.2 - 1 - 2 743 75

View Full Product Details