Assay Design



Gene Transcripts

Gene Symbol
ARHGAP40
Entrez Gene ID
419168
Gene Name
Rho GTPase activating protein 40
Gene Aliases
C20orf95
Chromosome Location
Chr.20: 3694821 - 3733103 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145016.1 NP_001138488.1 9 - 10 1324 89
XM_015296352.2 XP_015151838.1 9 - 10 1574 89
XM_015296353.2 XP_015151839.1 9 - 10 1574 89
XM_015296354.2 XP_015151840.1 9 - 10 1405 89
GenBank mRNA BX934665.2 - 6 - 7 707 89

View Full Product Details