Assay Design



Gene Transcripts

Gene Symbol
LOC416755
Entrez Gene ID
416755
Gene Name
uncharacterized LOC416755
Gene Aliases
-
Chromosome Location
Chr.14: 14193054 - 14236004 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015294773.2 XP_015150259.1 3 - 4 1003 77
XM_015294777.2 XP_015150263.1 3 - 4 1003 77

View Full Product Details