Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_004941776.3 | XP_004941833.1 | 1 - 2 | 260 | 100 |
XM_004941778.3 | XP_004941835.1 | 1 - 2 | 261 | 100 | |
XM_015287662.2 | XP_015143148.1 | 1 - 2 | 261 | 100 | |
GenBank mRNA | BX935451.2 | - | 1 - 2 | 256 | 100 |