Assay Design



Gene Transcripts

Gene Symbol
ARL5B
Entrez Gene ID
420513
Gene Name
ADP ribosylation factor like GTPase 5B
Gene Aliases
ARL8
Chromosome Location
Chr.2: 19078341 - 19094395 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143969.1 NP_001137441.1 1 - 2 118 67
GenBank mRNA CR385805.1 - 1 - 2 118 67

View Full Product Details