Assay Design



Gene Transcripts

Gene Symbol
TRUB1
Entrez Gene ID
423914
Gene Name
TruB pseudouridine synthase family member 1
Gene Aliases
-
Chromosome Location
Chr.6: 29011106 - 29039515 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015288931.2 XP_015144417.1 1 - 2 149 97
XM_421776.6 XP_421776.1 1 - 2 183 97
GenBank mRNA CR387253.1 - 1 - 2 133 97
CR389445.1 - 1 - 2 136 97

View Full Product Details