Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_204348.3 | NP_989679.3 | 2 - 3 | 318 | 65 |
XM_015281203.2 | XP_015136689.1 | 6 - 7 | 962 | 65 | |
GenBank mRNA | AF051944.2 | - | 2 - 3 | 284 | 65 |
BX932230.1 | - | 2 - 3 | 318 | 65 |