Assay Design



Gene Transcripts

Gene Symbol
WNT8B
Entrez Gene ID
428953
Gene Name
Wnt family member 8B
Gene Aliases
-
Chromosome Location
Chr.6: 18281587 - 18303981 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025151997.1 XP_025007765.1 2 - 3 14096 69
XM_025151998.1 XP_025007766.1 3 - 4 13559 69
GenBank mRNA AY753292.1 - 3 - 4 409 69

View Full Product Details