Assay Design



Gene Transcripts

Gene Symbol
TRAPPC6A
Entrez Gene ID
423337
Gene Name
trafficking protein particle complex 6A
Gene Aliases
TRAPPC6B
Chromosome Location
Chr.5: 37808851 - 37812670 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252160.1 NP_001239089.1 1 - 2 153 95
GenBank mRNA BU398441.1 - 1 - 2 158 95
BX932658.1 - 1 - 2 158 95
BX932692.1 - 1 - 2 257 95
BX935910.2 - 1 - 2 99 95

View Full Product Details