Assay Design
Gene Transcripts
- Gene Symbol
- MB21D2
- Entrez Gene ID
-
424905
- Gene Name
- Mab-21 domain containing 2
- Gene Aliases
-
C3orf59
- Chromosome Location
-
Chr.9: 13421714 - 13472199 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_422716.6 |
XP_422716.3 |
1 - 2 |
225 |
68 |
GenBank mRNA |
CR386427.1 |
- |
1 - 2 |
152 |
68 |
View Full Product Details