Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015289212.2 | XP_015144698.1 | 3 - 4 | 638 | 68 |
XM_025155075.1 | XP_025010843.1 | 2 - 3 | 376 | 68 | |
XM_416217.5 | XP_416217.3 | 2 - 3 | 557 | 68 | |
GenBank mRNA | CR406879.1 | - | 2 - 3 | 538 | 68 |