Assay Design



Gene Transcripts

Gene Symbol
NFAM1
Entrez Gene ID
417979
Gene Name
NFAT activating protein with ITAM motif 1
Gene Aliases
-
Chromosome Location
Chr.1: 49248294 - 49268747 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015289212.2 XP_015144698.1 5 - 6 849 57
XM_025155075.1 XP_025010843.1 4 - 5 587 57
XM_416217.5 XP_416217.3 4 - 5 768 57
GenBank mRNA CR406879.1 - 4 - 5 749 57

View Full Product Details