Assay Design



Gene Transcripts

Gene Symbol
BNIP3
Entrez Gene ID
423971
Gene Name
BCL2 interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.6: 35758032 - 35763615 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_421829.6 XP_421829.3 4 - 5 469 85
GenBank mRNA BX931008.2 - 4 - 5 463 85
BX932000.2 - 4 - 5 430 85
BX933976.1 - 4 - 5 408 85
BX933995.1 - 4 - 5 561 85
BX935850.2 - 4 - 5 438 85
CR407493.1 - 4 - 5 466 85

View Full Product Details