Assay Design



Gene Transcripts

Gene Symbol
SLC5A7
Entrez Gene ID
414870
Gene Name
solute carrier family 5 member 7
Gene Aliases
-
Chromosome Location
Chr.1: 137342523 - 137368789 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001763.1 NP_001001763.1 7 - 8 1118 72
XM_015277834.2 XP_015133320.1 8 - 9 1818 72
GenBank mRNA AJ511267.1 - 5 - 6 689 72

View Full Product Details