Assay Design
Gene Transcripts
- Gene Symbol
- PIH1D3
- Entrez Gene ID
-
422184
- Gene Name
- PIH1 domain containing 3
- Gene Aliases
-
C4HXORF41, CXorf41, NYSAR97
- Chromosome Location
-
Chr.4: 1905206 - 1913527 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_420180.6 |
XP_420180.3 |
5 - 6 |
510 |
72 |
GenBank mRNA |
BX934346.2 |
- |
5 - 6 |
494 |
72 |
View Full Product Details