Assay Design



Gene Transcripts

Gene Symbol
MIS18A
Entrez Gene ID
418494
Gene Name
MIS18 kinetochore protein A
Gene Aliases
C21orf45
Chromosome Location
Chr.1: 106218569 - 106223716 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_416702.6 XP_416702.1 1 - 2 73
GenBank mRNA CR353053.1 - 1 - 2 273 73
CR354097.1 - 1 - 2 251 73
CR391444.1 - 1 - 2 276 73

View Full Product Details