Assay Design



Gene Transcripts

Gene Symbol
VMO1
Entrez Gene ID
418974
Gene Name
vitelline membrane outer layer 1 homolog
Gene Aliases
ERGA6350, PRO21055, VMOI
Chromosome Location
Chr.1: 182061433 - 182065423 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167761.1 NP_001161233.1 1 - 2 183 84
GenBank mRNA D26093.1 - 1 - 2 184 84

View Full Product Details