Assay Design



Gene Transcripts

Gene Symbol
PTGS2
Entrez Gene ID
396451
Gene Name
prostaglandin-endoperoxide synthase 2
Gene Aliases
CEF147, PGHS2, PHSII
Chromosome Location
Chr.8: 10714136 - 10722126 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167718.1 NP_001161190.1 9 - 10 1908 123
NM_001167719.1 NP_001161191.1 9 - 10 1479 123
GenBank mRNA M64990.1 - 9 - 10 2004 123

View Full Product Details