Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190896.1 | NP_001177825.1 | 3 - 4 | 398 | 153 |
XM_015286159.2 | XP_015141645.2 | 3 - 4 | 320 | 153 | |
GenBank mRNA | BX930179.1 | - | 3 - 4 | 398 | 153 |
BX930378.2 | - | 3 - 4 | 320 | 153 |