Assay Design



Gene Transcripts

Gene Symbol
DNAJC24
Entrez Gene ID
421603
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Gene Aliases
DPH4, ZCSL3
Chromosome Location
Chr.5: 5240479 - 5272375 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190896.1 NP_001177825.1 4 - 5 464 110
XM_015286159.2 XP_015141645.2 4 - 5 386 110
GenBank mRNA BX930179.1 - 4 - 5 464 110
BX930378.2 - 4 - 5 386 110

View Full Product Details