Assay Design
Gene Transcripts
- Gene Symbol
- PUDP
- Entrez Gene ID
-
418655
- Gene Name
- pseudouridine 5'-phosphatase
- Gene Aliases
-
HDHD1, HDHD1A
- Chromosome Location
-
Chr.1: 127826522 - 127896240 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_416851.6 |
XP_416851.3 |
2 - 3 |
356 |
149 |
GenBank mRNA |
BX932630.1 |
- |
2 - 3 |
344 |
149 |
View Full Product Details