Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001305.3 | NP_001001305.1 | 3 - 3 | 1548 | 102 |
XM_015295121.2 | XP_015150607.1 | 3 - 3 | 2730 | 102 | |
XM_015295122.2 | XP_015150608.1 | 4 - 4 | 2460 | 102 | |
XM_025141547.1 | XP_024997315.1 | 4 - 4 | 1678 | 102 | |
GenBank mRNA | BX933423.1 | - | 1 - 1 | 56 | 102 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003642337.4 | XP_003642385.1 | 5 - 5 | 2078 | 102 |
XM_004946111.3 | XP_004946168.1 | 4 - 4 | 1949 | 102 |