Assay Design



Gene Transcripts

Gene Symbol
MRPL14
Entrez Gene ID
421443
Gene Name
mitochondrial ribosomal protein L14
Gene Aliases
-
Chromosome Location
Chr.3: 30494315 - 30503120 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025149086.1 XP_025004854.1 2 - 3 149 116
GenBank mRNA BX934464.1 - 2 - 3 132 116
BX935427.1 - 2 - 3 170 116

View Full Product Details