Assay Design



Gene Transcripts

Gene Symbol
CETN2
Entrez Gene ID
422668
Gene Name
centrin 2
Gene Aliases
CETN1
Chromosome Location
Chr.4: 53633118 - 53638015 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277920.1 NP_001264849.1 1 - 2 64 73
GenBank mRNA BX930994.2 - 1 - 2 64 73
BX935748.1 - 1 - 2 66 73

View Full Product Details