Assay Design



Gene Transcripts

Gene Symbol
PRKX
Entrez Gene ID
418656
Gene Name
protein kinase, X-linked
Gene Aliases
-
Chromosome Location
Chr.1: 129430051 - 129485175 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015277728.2 XP_015133214.1 5 - 6 1139 74
GenBank mRNA CR354088.1 - 5 - 6 736 74

View Full Product Details