Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_004943567.3 | XP_004943624.1 | 2 - 3 | 2027 | 109 |
XM_015291844.1 | XP_015147330.1 | 3 - 4 | 2182 | 109 | |
XM_025153460.1 | XP_025009228.1 | 2 - 3 | 2027 | 109 | |
XM_025153461.1 | XP_025009229.1 | 2 - 3 | 2023 | 109 | |
XM_422836.6 | XP_422836.2 | 2 - 3 | 418 | 109 | |
GenBank mRNA | BX929777.1 | - | 2 - 3 | 374 | 109 |
BX931204.1 | - | 2 - 3 | 408 | 109 |