Assay Design
Gene Transcripts
- Gene Symbol
- GATC
- Entrez Gene ID
-
416976
- Gene Name
- glutamyl-tRNA amidotransferase subunit C
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 9576606 - 9578696 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_415269.6 |
XP_415269.3 |
2 - 3 |
309 |
79 |
GenBank mRNA |
BX930114.1 |
- |
2 - 3 |
280 |
79 |
View Full Product Details