Assay Design



Gene Transcripts

Gene Symbol
NIM1K
Entrez Gene ID
768482
Gene Name
NIM1 serine/threonine protein kinase
Gene Aliases
NIM1, NIM1KZ
Chromosome Location
Chr.Z: 13642221 - 13655186 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015272679.2 XP_015128165.1 2 - 3 1655 121
GenBank mRNA CR406079.1 - 1 - 2 437 121

View Full Product Details