Assay Design
Gene Transcripts
- Gene Symbol
- NIM1K
- Entrez Gene ID
-
768482
- Gene Name
- NIM1 serine/threonine protein kinase
- Gene Aliases
-
NIM1, NIM1KZ
- Chromosome Location
-
Chr.Z: 13642221 - 13655186 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CR406079.1 |
- |
2 - 3 |
550 |
118 |
View Full Product Details