Assay Design



Gene Transcripts

Gene Symbol
TPRG1L
Entrez Gene ID
419384
Gene Name
tumor protein p63 regulated 1-like
Gene Aliases
FAM79A
Chromosome Location
Chr.21: 992782 - 996073 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015297097.2 XP_015152583.1 4 - 5 641 78
GenBank mRNA BX933400.2 - 2 - 3 188 78
CR406495.1 - 4 - 5 631 78

View Full Product Details