Assay Design



Gene Transcripts

Gene Symbol
KLHL36
Entrez Gene ID
415816
Gene Name
kelch like family member 36
Gene Aliases
C16orf44
Chromosome Location
Chr.11: 16836374 - 16853710 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004944268.3 XP_004944325.1 3 - 4 1315 67
XM_004944269.3 XP_004944326.1 4 - 5 1707 67
XM_004944270.3 XP_004944327.1 3 - 4 1326 67
XM_015292625.2 XP_015148111.1 4 - 5 1704 67
XM_015292626.2 XP_015148112.1 2 - 3 1548 67
XM_025154330.1 XP_025010098.1 3 - 4 1299 67
XM_025154331.1 XP_025010099.1 4 - 5 1680 67
XM_025154332.1 XP_025010100.1 4 - 5 1677 67
XM_025154333.1 XP_025010101.1 3 - 4 1396 67
XM_414178.6 XP_414178.3 3 - 4 1288 67
GenBank mRNA CR523083.1 - 1 - 2 553 67

View Full Product Details