Assay Design



Gene Transcripts

Gene Symbol
GSTK1
Entrez Gene ID
418302
Gene Name
glutathione S-transferase kappa 1
Gene Aliases
-
Chromosome Location
Chr.1: 78425551 - 78436579 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198649.1 NP_001185578.1 7 - 8 682 140
XM_025149199.1 XP_025004967.1 6 - 7 678 140
GenBank mRNA BX930797.1 - 7 - 8 646 140
BX931011.2 - 7 - 8 682 140
BX934516.2 - 7 - 8 648 140
CR387708.1 - 6 - 7 600 140

View Full Product Details